Anti-ST6GALNAC1 Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB69066-50EA 11400 NOK
ABCAAB69066-50
Anti-ST6GALNAC1 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ST6GALNAC1 (ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR