Anti-GOLPH3 Mouse polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB69171-50EA 11475 NOK
ABCAAB69171-50
Anti-GOLPH3 Mouse polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: GOLPH3 (golgi phosphoprotein 3 (coat-protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human