Anti-ZC3H10 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB69209-50EA 10900 NOK
ABCAAB69209-50
Anti-ZC3H10 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ZC3H10 (zinc finger CCCH-type containing 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human