Anti-Legionella pneumophila Mouse Monoclonal Antibody [clone: 142]

Leverandør: Abcam
ABCAAB69239-100EA 8350 NOK
ABCAAB69239-100
Anti-Legionella pneumophila Mouse Monoclonal Antibody [clone: 142]
Antistoffer
Type: Primary
Antigen: Legionella pneumophila
Clonality: monoclonal
Clone: 142
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR