Anti-Salmonella Rabbit Polyclonal Antibody (Biotin)
Leverandør: Abcam
ABCAAB69255-500EA 11400 NOK
ABCAAB69255-500
Anti-Salmonella Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Salmonella
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå