Anti-GPT2 Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB69279-50EA 11400 NOK
ABCAAB69279-50
Anti-GPT2 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: GPT2 (glutamic pyruvate transaminase (alanine aminotransferase) 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR