Anti-MED9 Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB69323-50EA 8050 NOK
ABCAAB69323-50
Anti-MED9 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MED9 (mediator complex subunit 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR