Anti-CHCHD3 Mouse polyclonal antibody unconjugated

ABCAAB69328-50EA 7725 NOK
ABCAAB69328-50
Anti-CHCHD3 Mouse polyclonal antibody unconjugated
Type: Primary
Antigen: CHCHD3 (coiled-coil-helix-coiled-coil-helix domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR