Anti-CYP4V2 Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB69392-50EA 10900 NOK
ABCAAB69392-50
Anti-CYP4V2 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CYP4V2 (cytochrome P450, family 4, subfamily V, polypeptide 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR