Anti-ERCC1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB69398-100EA 11150 NOK
ABCAAB69398-100
Anti-ERCC1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ERCC1 (excision repair cross-complementation group 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR