Anti-DFFB Rabbit polyclonal antibody unconjugated

ABCAAB69438-100EA 7250 NOK
ABCAAB69438-100
Anti-DFFB Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: DFFB (DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR