Anti-NUP43 Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB69447-50EA 10900 NOK
ABCAAB69447-50
Anti-NUP43 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NUP43 (nucleoporin 43kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR