Anti-VEGF 165A Mouse Monoclonal Antibody [clone: 6B7]
Leverandør: Abcam
ABCAAB69479-100EA 11650 NOK
ABCAAB69479-100
Anti-VEGF 165A Mouse Monoclonal Antibody [clone: 6B7]
Antistoffer
Type: Primary
Antigen: VEGF 165A
Clonality: monoclonal
Clone: 6B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse