Anti-GANAB Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB69489-100EA 11300 NOK
ABCAAB69489-100
Anti-GANAB Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: GANAB (glucosidase, alpha; neutral AB)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå