Vårt Oslo-kontor flytter den 25. mars 2019 til Brynsalleen 4, 0667 Oslo
Anti-PDGFRB Mouse monoclonal antibody unconjugated [clone: 42G12]
Leverandør: Abcam
Type: Primary
Antigen: PDGFRB (platelet-derived growth factor receptor, beta polypeptide)
Clonality: monoclonal
Clone: 42G12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
BESTILL
Utvid alle / Skjul alle
Str Fp Art.nr Antall
100 µg 100 µG ABCAAB69506-100
Lag barcode etiketter Barcode etikett format: 
Lag PDF katalog
SPESIFIKASJONER
Antigen symbol PDGFRB
Antigen name Platelet-derived growth factor receptor beta
Reaktivitet Rat, Mouse, Human
Konjugasjon Unconjugated
Klonalitet Monoclonal
antistofftype Primary
Klon 42G12
Vert Mouse
Isotype IgG1
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Lagringstemp Cool
Frakttemp. Ice
Legg inn detaljer
Velg varen hvor du søker etter sertifikat
Legg inn batchnummer. Legg inn "X" hvis du ikke har informasjonen for VWR Chemicals. Feltet er obligatorisk
Skriv inn serifikatdetaljer
Skriv inn lot nummer:
Velg fra de siste batch / lot tallene:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivelse Art.nr. Availability Enhet Din pris Price Per Antall