Anti-Caspase 9 p10 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB69514-100EA 11475 NOK
ABCAAB69514-100
Anti-Caspase 9 p10 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Caspase 9 p10
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå