Anti-MAP2K5 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB69537-100EA 8350 NOK
ABCAAB69537-100
Anti-MAP2K5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MAP2K5 (mitogen-activated protein kinase kinase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR