Anti-CASP7 Mouse monoclonal antibody unconjugated [clone: 7-1-11]

Leverandør: Abcam
ABCAAB69540-100EA 12275 NOK
ABCAAB69540-100
Anti-CASP7 Mouse monoclonal antibody unconjugated [clone: 7-1-11]
Antistoffer
Type: Primary
Antigen: CASP7 (caspase 7, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: 40725
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR