Anti-HIPK4 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB69565-100EA 10900 NOK
ABCAAB69565-100
Anti-HIPK4 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HIPK4 (homeodomain interacting protein kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human