Anti-Adenylate cyclase 1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB69597-100EA 11650 NOK
ABCAAB69597-100
Anti-Adenylate cyclase 1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Adenylate cyclase 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR