Anti-STK31 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB69678-50EA 10900 NOK
ABCAAB69678-50
Anti-STK31 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: STK31 (serine/threonine kinase 31)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human