Anti-EXOSC5 Mouse polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB69699-50EA 11725 NOK
ABCAAB69699-50
Anti-EXOSC5 Mouse polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: EXOSC5 (exosome component 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse