Anti-GLI3 Rabbit polyclonal antibody unconjugated

ABCAAB69838-100EA 7600 NOK
ABCAAB69838-100
Anti-GLI3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: GLI3 (GLI family zinc finger 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR