Anti-TMC8 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB69859-100EA 9475 NOK
ABCAAB69859-100
Anti-TMC8 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TMC8 (transmembrane channel-like 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR