Anti-PRPF4 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB69878-50EA 11475 NOK
ABCAAB69878-50
Anti-PRPF4 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PRPF4 (pre-mRNA processing factor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human