Anti-MAP4K4 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB69885-100EA 11475 NOK
ABCAAB69885-100
Anti-MAP4K4 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MAP4K4 (mitogen-activated protein kinase kinase kinase kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human