Anti-NANOS3 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70001-50EA 11900 NOK
ABCAAB70001-50
Anti-NANOS3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NANOS3 (nanos homolog 3 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat