Anti-HTN1 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB70024-50EA 11400 NOK
ABCAAB70024-50
Anti-HTN1 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HTN1 (histatin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human