Anti-GBP4 Mouse polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB70058-50EA 10900 NOK
ABCAAB70058-50
Anti-GBP4 Mouse polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: GBP4 (guanylate binding protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR