Anti-KPNA2 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB70160-100EA 11725 NOK
ABCAAB70160-100
Anti-KPNA2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KPNA2 (karyopherin alpha 2 (RAG cohort 1, importin alpha 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR