Anti-HUWE1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB70161-100EA 11975 NOK
ABCAAB70161-100
Anti-HUWE1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HUWE1 (HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR