Anti-PFKFB2 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB70175-100EA 12050 NOK
ABCAAB70175-100
Anti-PFKFB2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PFKFB2 (6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR