Anti-RPS6 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70227-100EA 12050 NOK
ABCAAB70227-100
Anti-RPS6 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RPS6 (ribosomal protein S6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse