Anti-DIS Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB70243-100EA 10900 NOK
ABCAAB70243-100
Anti-DIS Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: DIS
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human