Anti-RIF1 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70254-100EA 11475 NOK
ABCAAB70254-100
Anti-RIF1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: RIF1 (RAP1 interacting factor homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human