Anti-INPPL1 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70267-100EA 11775 NOK
ABCAAB70267-100
Anti-INPPL1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: INPPL1 (inositol polyphosphate phosphatase-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse