Anti-DHX36 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB70269-50EA 11975 NOK
ABCAAB70269-50
Anti-DHX36 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: DHX36 (DEAH (Asp-Glu-Ala-His) box polypeptide 36)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR