Anti-MSH2 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB70270-100EA 12200 NOK
ABCAAB70270-100
Anti-MSH2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MSH2 (mutS homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR