Anti-CTCF Rabbit polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB70303-100EA 12275 NOK
ABCAAB70303-100
Anti-CTCF Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: CTCF (CCCTC-binding factor (zinc finger protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse