Anti-UBR5 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70311-100EA 11725 NOK
ABCAAB70311-100
Anti-UBR5 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: UBR5 (ubiquitin protein ligase E3 component n-recognin 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse