Anti-FOXO3 Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB70314-100EA 11725 NOK
ABCAAB70314-100
Anti-FOXO3 Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: FOXO3 (forkhead box O3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR