Anti-SEC23IP Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70339-100EA 11400 NOK
ABCAAB70339-100
Anti-SEC23IP Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SEC23IP (SEC23 interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human