Anti-NCAPD3 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70349-100EA 11650 NOK
ABCAAB70349-100
Anti-NCAPD3 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: NCAPD3 (non-SMC condensin II complex, subunit D3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human