Anti-PCK1 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70358-100EA 11900 NOK
ABCAAB70358-100
Anti-PCK1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PCK1 (phosphoenolpyruvate carboxykinase 1 (soluble))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå