Anti-MCM2 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70371-100EA 11775 NOK
ABCAAB70371-100
Anti-MCM2 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MCM2 (minichromosome maintenance complex component 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human