Anti-CMTR1 Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB70386-100EA 10900 NOK
ABCAAB70386-100
Anti-CMTR1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CMTR1 (cap methyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human