Anti-P4HB Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB70415-50EA 8700 NOK
ABCAAB70415-50
Anti-P4HB Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: P4HB (prolyl 4-hydroxylase, beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR