Anti-PAXIP1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB70434-100EA 11475 NOK
ABCAAB70434-100
Anti-PAXIP1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PAXIP1 (PAX interacting (with transcription-activation domain) protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR