Anti-CNTROB Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB70448-50EA 11650 NOK
ABCAAB70448-50
Anti-CNTROB Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CNTROB (centrobin, centrosomal BRCA2 interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human