Anti-BCL11B Rabbit polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB70452-100EA 8700 NOK
ABCAAB70452-100
Anti-BCL11B Rabbit polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: BCL11B (B-cell CLL/lymphoma 11B (zinc finger protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR